missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TheraPure™ RNA Polymerases

Catalog No. FEREP011T1
Change view
Click to view available options
Polymerase:
T7 RNA Polymerase
Concentration:
200 U/μL
Quantity:
20,000 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Polymerase Concentration Quantity
FEREP011T1 T7 RNA Polymerase 200 U/μL 20,000 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEREP011T1 Supplier Thermo Scientific™ Supplier No. EP011T1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific TheraPure in vitro transcription (IVT) enzymes are designed for advanced research and early development of mRNA therapeutics and vaccines.

Thermo Scientific TheraPure in vitro transcription (IVT) enzymes are designed for advanced research and early development of mRNA therapeutics and vaccines. These products offer full workflow solutions from template preparation to RNA synthesis and are recommended for process research and optimization, proof of concept, and preclinical studies.

The high purity of TheraPure IVT enzymes is ensured by precise quality testing that includes confirmation that that the enzymes are nuclease- and endotoxin-free. In addition, the manufacturing process of TheraPure products follows ISO9001 and ISO13485 standards.

Features

  • Convenient stand-alone enzymes for easy workflow optimization
  • Animal-origin-free (AOF) raw materials used in manufacturing
  • No risk of prion or animal virus contamination
  • Reproducible and consistent results with validated quality testing
  • Regulatory compliant—β-lactam-free manufacturing technology
  • Suitable for translational research
  • Facilitates transition to GMP during mRNA vaccine development process
  • Scalable and available as OEM product

The exceptional quality of TheraPure products is supported by:

  • Certificates of analysis
  • Certificates of origin
  • TSE/BSE statement

Specifications

Polymerase T7 RNA Polymerase
Product Line TheraPure
Promoter T7
For Use With (Application) In vitro Transcription
Quantity 20,000 U
Concentration 200 U/μL

For Research Use Only.