missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TheraPure™ Nucleotides

Catalog No. FERR044T1
Change view
Click to view available options
Product Type:
ATP Solution, Sodium Solution
ATP Solution, Tris Solution
CTP Solution, Sodium Solution
CTP Solution, Tris Solution
GTP Solution, Sodium Solution
GTP Solution, Tris Solution
N1-Methylpseudo-UTP, Sodium Solution
UTP Solution, Sodium Solution
UTP Solution, Tris Solution
Concentration:
100 mM
200 mM
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Concentration
FERR044T1 ATP Solution, Sodium Solution 100 mM
FERR00044T1 ATP Solution, Tris Solution 200 mM
FERR045T1 CTP Solution, Sodium Solution 100 mM
FERR00045T1 CTP Solution, Tris Solution 200 mM
FERR046T1 GTP Solution, Sodium Solution 100 mM
FERR00046T1 GTP Solution, Tris Solution 200 mM
FERR047T1 UTP Solution, Sodium Solution 100 mM
FERR00047T1 UTP Solution, Tris Solution 200 mM
FERR0491T1 N1-Methylpseudo-UTP, Sodium Solution 100 mM
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. FERR044T1 Supplier Thermo Scientific™ Supplier No. R044T1

Thermo Scientific TheraPure in vitro transcription (IVT) enzymes are designed for advanced research and early development of mRNA therapeutics and vaccines.

Thermo Scientific TheraPure in vitro transcription (IVT) enzymes are designed for advanced research and early development of mRNA therapeutics and vaccines. These products offer full workflow solutions from template preparation to RNA synthesis and are recommended for process research and optimization, proof of concept, and preclinical studies.

The high purity of TheraPure IVT enzymes is ensured by precise quality testing that includes confirmation that that the enzymes are nuclease- and endotoxin-free. In addition, the manufacturing process of TheraPure products follows ISO9001 and ISO13485 standards.

Features

  • Convenient stand-alone enzymes for easy workflow optimization
  • Animal-origin-free (AOF) raw materials used in manufacturing
  • No risk of prion or animal virus contamination
  • Reproducible and consistent results with validated quality testing
  • Regulatory compliant—β-lactam-free manufacturing technology
  • Suitable for translational research
  • Facilitates transition to GMP during mRNA vaccine development process
  • Scalable and available as OEM product

The exceptional quality of TheraPure products is supported by:

  • Certificates of analysis
  • Certificates of origin
  • TSE/BSE statement

Specifications

Product Type ATP Solution, Sodium Solution
Content And Storage Store at -20°C.
Shipping Condition Dry Ice
Concentration 100 mM
Quantity 0.2 mL
Product Line TheraPure
Color Colorless
For Use With (Application) In vitro Transcription

For Research Use Only.