missing translation for 'onlineSavingsMsg'
Learn More

Therapak™ TearTech™ 25-Hole Foam Rack

Catalog No. 22130076
Change view
Click to view available options
Type:
25 Hole Foam Rack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
22130076 25 Hole Foam Rack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22130076 Supplier Therapak™ Supplier No. 68825

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Versatile tube transport system that provides cushioning and protection for blood tubes, urinalysis tubes and many other commonly transported test tubes

  • Accommodates standard tube sizes from 13 × 75mm to 16 × 100mm
  • Overall foam block size is 150 × 150 × 100mm
  • Die-cut construction permits tearing away variable grid sizes from 1 to 25
  • Provides excellent protection for tubes during transport
  • Combine with boxes, insulated shippers and bags for specimen transport

Specifications

Quantity Up to 25 tubes (12-16mm dia.) with length of 100mm
Type 25 Hole Foam Rack
Dimensions (LxWxH) 150 x 150 x 100 mm
Capacity 25 Tubes
Material Foam