Learn More
Therapak™ Single Specimen Collection Kits
Single Specimen Collection Kits can be used in conjunction with on-site testing products or remote site collection when specimen is transported to a laboratory for analysis.
Supplier:  Therapak™ TR40005
 View more versions of this product
Catalog No. 22131554
Only null left
Description
Description
Single Specimen Collection Kits can be used in conjunction with on-site testing products or remote site collection when specimen is transported to a laboratory for analysis. Each kit contains either a 180 mL beaker style collection cup with pour spout or a leak-resistant 90 mL sterile container with cap and integrity seal. The transport bag features two sealable, tamper evident pockets and comes with absorbent material. The individually bagged kits are available in various configurations.
Specifications
Chemical Identifiers
Urine Specimen Container
For drugs of abuse testing, on-site testing or remote site collection
90 mL
Sterile
3 oz.
Specifications
Urine Specimen Container
For drugs of abuse testing, on-site testing or remote site collection
3 oz.
Sterile
Temp. Strip, Evidence Seal, Bag
90 mL
Product Suggestions
Product Suggestions