missing translation for 'onlineSavingsMsg'
Learn More

Therapak™ Plastic Duramark™ Lock Boxes

Catalog No. 22131452
Change view
Click to view available options
Includes:
Lock Box
Lock Box with Over Door Strap
Lock Box with Under Door Strap
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
22131452 Lock Box
22130206 Lock Box with Over Door Strap
22131450 Lock Box with Under Door Strap
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22131452 Supplier Therapak™ Supplier No. 36745

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High impact polystyrene construction

  • Made for hanging on the door using the optional straps
  • Provides the timesaving convenience of a viewing window and a hinged door that opens down for ease of use
  • Comes with lock and two matching keys for adding security
  • Dimensions: 13.5L x 10.25W x 6.75 in. H
  • Standard color: Off-white
  • Standard printing: Reflex blue text, red biohazard symbol on front side only
  • Optional straps: Under the door, over the door
  • Customization: Any deviation from the standard stock configuration, which may include changes in color, size, design features, closure and print

Specifications

Includes Lock Box
Color Off-White
Dimensions (LxWxH) 13.5 x 10.25 x 6.75 in.
Material High-impact Polystyrene