missing translation for 'onlineSavingsMsg'
Learn More

Therapak™ Metal Duramark™ Lock Boxes

Specifications

Material 24 gauge cold rolled steel construction
View More Specs

Products 2
Catalog Number Mfr. No. Includes Price Quantity & Availability  
Catalog Number Mfr. No. Includes Price Quantity & Availability  
22130207
View Documents Promotion Details
Therapak™
36507
Metal Lock Box + Over Door Strap N/A
Only null left
 
22131453
View Documents Promotion Details
Therapak™
36746
Metal Lock Box N/A
Only null left
 
Description

Description

  • 24 Gauge cold rolled steel construction
  • Painted with powder coat finish for excellent strength and durability
  • Insulated on inside with foam assisting preserving the integrity of the specimens
  • With hinged front door that rotates down 90° for convenient access and use as temporary tray
  • Dimensions: 14 × 10 1/2 × 7 in.
  • Supplied with lock and two matching keys for security
  • Standard color: White
  • Standard printing: Reflex blue text, red biohazard symbol on front side only
  • Optional straps: Under the door, over the door
  • Customization: Any deviation from the standard stock configuration, which may include changes in color, size, design features, closure and print

Specifications

Specifications

24 gauge cold rolled steel construction
Videos
SDS
Documents

Documents