Learn More
Therapak™ Gel Pack Protective Barrier Transport Bag
Insulate specimen samples requiring ambient to cool environment during transit
Supplier: Therapak™ TR15515
Description
Just add cold water to designated line to form gel.
- Lightweight, compact alternative to standard insulating coolants
- 6 × 9 in. (15.2 × 22.9cm) zip style bag
- Pillow pack materials are nontoxic and noncarcinogenic
- Bags store flat and are virtually weightless until activated
Specifications
| 9 x 6 in. (22.9 x 15.2cm) | |
| 6 in. | |
| 15.2 cm |
| 9 in. | |
| 22.9 cm |