missing translation for 'onlineSavingsMsg'
Learn More

Therapak™ Frozen Specimen Label

Catalog No. 22130119
Change view
Click to view available options
Size (Metric):
5.08 x 2.22 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size (Metric)
22130119 5.08 x 2.22 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22130119 Supplier Therapak™ Supplier No. 54609

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Convenient marking label for frozen content identification

  • 500 labels per roll

Specifications

Certifications/Compliance DOT, IATA
Format Label
Product Type Frozen Specimen Label
For Use With (Application) Convenient marking label for frozen content identification
Quantity 1000/Roll
Size (English) 2 x 0.875 in.
Size (Metric) 5.08 x 2.22 cm
Length (English) 2 in.
Width (English) 0.875 in.