Learn More
Therapak™ Exempt Human Specimen (EHS) Ambient Shipping Systems
Description
- Use to ship patient specimens with a minimal likelihood of pathogens, including blood or urine for cholesterol levels; hormone levels; prostate specific antigens; tests to monitor organ function (heart, liver, kidney); tests conducted for insurance or employment; pregnancy; biopsies for cancer; drug or alcohol presence testing
- Ambient Systems for tubes, containers, and bags include a transport box, Exempt Human Specimen label, specimen bag, absorbent material, biohazard label, cushioning material, and packing instructions
- Bulk Ambient TearTech™ Shipper includes 50 pop-up-style TearTech boxes, 100 zip style biohazard transport bags (two pocket bag for sample and requisition form), 100 Aqui-Pak™ segmented absorbent pouches (each Aqui-Pak holds up to six standard blood or urine tubes), 50 biohazard labels and 50 Exempt Human Specimen labels
Compliance
All systems comply with DOT and IATA regulations for ground and air shipments of specimens.
Specifications
Specifications
| Quantity | 1 Ea. |
| Certifications/Compliance | All systems comply with IATA regulations for ground and air shipments of specimens. |
| Applications | For standard tube or larger specimen containers (30mL to 60mL) of liquid specimen |
| Capacity (English) | Holds 1 primary container |