missing translation for 'onlineSavingsMsg'
Learn More

Therapak™ Exempt Human Specimen (EHS) Ambient Shipping Systems

Catalog No. 22130429
Change view
Click to view available options
Capacity (English):
Holds 1 primary container
Holds 10- 6 x 9 in. specimen bags
Holds 4 tubes
Holds 4- 6 x 9 in. specimen bags
Holds 6 tubes
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
22130429 Holds 10- 6 x 9 in. specimen bags
22130426 Holds 1 primary container
22130427 Holds 4 tubes
22130425 Holds 4- 6 x 9 in. specimen bags
22130428 Holds 6 tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 22130429 Supplier Therapak™ Supplier No. 56875

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For hospitals, hospital laboratories and reference laboratory testing

  • Use to ship patient specimens with a minimal likelihood of pathogens, including blood or urine for cholesterol levels; hormone levels; prostate specific antigens; tests to monitor organ function (heart, liver, kidney); tests conducted for insurance or employment; pregnancy; biopsies for cancer; drug or alcohol presence testing
  • Ambient Systems for tubes, containers, and bags include a transport box, Exempt Human Specimen label, specimen bag, absorbent material, biohazard label, cushioning material, and packing instructions
  • Bulk Ambient TearTech™ Shipper includes 50 pop-up-style TearTech boxes, 100 zip style biohazard transport bags (two pocket bag for sample and requisition form), 100 Aqui-Pak™ segmented absorbent pouches (each Aqui-Pak holds up to six standard blood or urine tubes), 50 biohazard labels and 50 Exempt Human Specimen labels

Compliance

All systems comply with DOT and IATA regulations for ground and air shipments of specimens.

Specifications

Quantity 1 Ea.
Includes 12 x 18 in. biohazard transport bag, 12 x 12 in. absorbent sheet; Holds ten 6 x 9 in. specimen transport bags
Certifications/Compliance All systems comply with IATA regulations for ground and air shipments of specimens.
Type Shipping Container
Capacity (English) Holds 10- 6 x 9 in. specimen bags
Dimensions (LxWxH) 11 x 10.5 x 4 in.
Capacity 10 x 6 x 9 in. Specimen Transport Bags
Material Corrugated Cardboard