missing translation for 'onlineSavingsMsg'
Learn More

Therapak™ Exempt Animal Specimen Labels

Catalog No. 22130066
Change view
Click to view available options
Size (Metric):
8.8 x 2.5 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size (Metric)
22130066 8.8 x 2.5 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22130066 Supplier Therapak™ Supplier No. 54751

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Complies with IATA regulations for shipping veterinary specimens

Exempt Animal Specimen category applies to shipment of animal specimens for which, through professional judgment, has been determined there is minimal likelihood that pathogens are present.

  • Easy-to-use roll format
  • Simply apply the self-adhesive label to the exterior of complaint packaging prior to shipping
  • 500 labels per roll

Specifications

Certifications/Compliance IATA
Adhesive Type Self-adhesive
Format Label
Product Type Exempt Animal Specimen Label
For Use With (Application) For shipping veterinary specimens
Quantity 500/Pk.
Size (English) 3.5 x 1 in.
Size (Metric) 8.8 x 2.5 cm
Length (English) 3.5 in.
Width (English) 1 in.