Learn More
Therapak™ EAS Frozen Shipper
Catalog No. 22130439
Change view
Click to view available options
Type:
Frozen Shipper
Catalog No. Type
22130439 Frozen Shipper
1 options
Catalog No. 22130439 Supplier Therapak™ Supplier No. TR56885
Includes
Specimen bag, Absorbent material, Exempt Animal Specimen label, Biohazard symbol label, and Cushioning material
Ideal for overnight shipments via common carriers such as UPS™ and FedEx™
The International Air Transport Association (IATA) Dangerous Goods Regulations (DGR) manual contains a category that was created for shipping animal specimens for which there is minimal likelihood that pathogens are present. The regulations cite examples of many routine laboratory tests that can now be transported as Exempt Animal Specimens (EAS). Examples of those tests are: blood or urine for cholesterol levels, hormone levels, prostate specific antigens (PSA), tests to monitor organ function (heart, liver, kidney), tests conducted for pregnancy and biopsies for cancer.
  • Standard small insulated cooler and box with Class 9 dry ice label
  • Add sample tubes and dry ice to ship
Meets following IATA Exempt Animal Specimen package requirements:
  • 100 × 100mm dimension on one side
  • Leakproof primary container
  • Leakproof secondary container
  • Absorbent between primary and secondary sufficient to absorb total primary container(s) contents
  • Outer packaging of adequate strength for its capacity
Order Info
Cooler L x W x H: 11 x 9 x 11 in. (exterior), 8 x 6 x 8 in. (interior)
Specifications
Includes Specimen bag, absorbent/cushioning material, Exempt Animal Specimen and biohazard labels
Type Frozen Shipper
Dimensions (LxWxH) 11 x 9 x 11 in.