Learn More
Therapak™ Duramark™ Thermoelectric Cooler/Warmer
Active solution for the transport of laboratory medical specimens
Supplier: Therapak™ 562060G
Description
- Use of 12V power plug in vehicle, each unit will cool to approximately 40°F lower than ambient temperature or heat to around 135°F
- Optional equipment includes battery saver switch which protects the car's battery from premature draining
- AC Power adapter allows the unit to be used in any 110/120 volt AC wall outlet
Order Info
Battery, adapter and power cords available separately (Cat. Nos. 22-131-495, 22-131-491 and 22-131-494).
Specifications
| 43.8 cm | |
| 40.6 cm | |
| 21.5 cm | |
| 16 x 8.5 x 17.25 in. | |
| 10 lb. | |
| Compact Cooler, 12V |
| 17.25 in. | |
| 16 in. | |
| 8.5 in. | |
| For transport of laboratory medical specimens | |
| 4.53 kg | |
| 17 L |