missing translation for 'onlineSavingsMsg'
Learn More

Therapak™ Duramark™ Fabric Lock Boxes

Available in standard small and large sizes

Supplier:  Therapak™ TR36501

 View more versions of this product

Catalog No. 22-131-426


Only null left

Description

Description

  • Soft sides insulated with 1/4 in. foam
  • Consists of zipper closure, mesh envelope to contain refrigerant packs, and an adjustable strap for ease of courier handling during pickup activity
  • Optional zipper lock includes two matching keys, and optional security strap, which is reinforced with a steel cord sewn into the strap, helps assure the sample will be safe and secure
Standard color: Royal Blue Regency
Standard printing: Silver text with red biohazard symbol on one side only
Options include: Zipper lock, security strap
Customization: Any deviation from the standard stock configuration, which may include changes in color, size, design features (i.e. number of pockets), closure (i.e., Zipper, Velcro, buckle, etc.) and print
Minimum customization order quantity is 24 bags.
Artwork creation is available.
Small Size Dimensions: 9 x 8 x 7 in. (22.86 x 20.3 x 17.7cm)
Large Size Dimensions: 11 x 9 x 7 in. (27.9 x 22.86 x 17.7cm)

Specifications

Specifications

Lock Box, Zipper Lock, Keys
Small
Fabric
Royal Blue Regency
With Red Biohazard Symbol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions