missing translation for 'onlineSavingsMsg'
Learn More

Therapak™ Dry Ice Label

Catalog No. 22130065
Change view
Click to view available options
Size (Metric):
13.9 x 13.9 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size (Metric)
22130065 13.9 x 13.9 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22130065 Supplier Therapak™ Supplier No. 54530

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Complies with DOT and IATA regulations for all dry ice shipments

  • Self-adhesive roll stock for peel and apply convenience
  • Preprinted with all required information—simply complete designated Shipper and Consignee name and address, amount of dry ice (in kg) included in package, and apply label to package

Compliance

Complies with DOT (49 CFR 173.217) and IATA (Packing Instructions 904) regulations.

Specifications

Certifications/Compliance DOT 49 CFR 173.217, IATA
Adhesive Type Self-adhesive
Format Label
Product Type Dry Ice Shipping Label
For Use With (Application) Used to transport biological specimens
Quantity 500/Pk.
Size (English) 5.5 x 5.5 in.
Size (Metric) 13.9 x 13.9 cm
Length (English) 5.5 in.
Width (English) 5.5 in.