Learn More
Therapak™ Drugs of Abuse Replenishment Supplies
Supplies for use with single- and split-specimen collection kits.
Supplier:  Therapak™ TR40008
 View more versions of this product
Catalog No. 22131560
Only null left
Description
Description
Specifications
Chemical Identifiers
Non-sterile
Single and split specimen collection kits in drugs of abuse testing
Specifications
Tamper Evident Security Seal
Non-sterile
Used for chain of custody procedures in drug testing, identify the presence of any tampering with the specimen containers or vials from the moment they are sealed until they are opened at the testing laboratory
500 Pk.
Acetate Film with Paper Backing
Red
Single and split specimen collection kits in drugs of abuse testing
Product Suggestions
Product Suggestions