Learn More
Therapak™ Drugs of Abuse Replenishment Supplies
Catalog No. 22131559
Change view
Click to view available options
Product Type:
Specimen Transport Bag
Tamper Evident Security Seal
Temperature Indicator Strip
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
22131559 Temperature Indicator Strip
22131560 Tamper Evident Security Seal
22131561 Specimen Transport Bag
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22131559 Supplier Therapak™ Supplier No. TR40006
Supplies for use with single- and split-specimen collection kits.
Specifications
Product Type Temperature Indicator Strip
Sterility Non-sterile
For Use With (Application) Indicate the temperature of the specimen within minutes after the initial collection
For Use With (Equipment) Single and split specimen collection kits in drugs of abuse testing
Quantity 100 Pk.
Temperature Range (English) 90 to 100°F
Temperature Range (Metric) 32 to 38°C