Learn More
Therapak™ Courier Totes
Temperature-sensitive insulation protects the safety and integrity of refrigerated and frozen specimens
Supplier: Therapak™ TR36510
Description
- Shoulder-strap specimen totes constructed of waterproof, scuff-resistant nylon
- Standard 2 in. foldover closure forms secure seal over lid
- Expandable pouch on back of tote holds additional ambient specimens; two document pockets secure requisitions and reports
- Adjustable dividers allow courier to accommodate various shipments
- Available in two sizes with molded, high R-value urethane or EPS foam insert
- Small tote in blue; standard tote in black preprinted with a 2 × 2 1/2 in. red international biohazard symbol
- Meet OSHA and DOT requirements for shipments of biological substances via ground courier transport
- replacement inserts available separately
Specifications
| Meets OSHA and DOT requirements for shipments of biological substances via ground courier transport | |
| Standard | |
| 45.7 x 22.8 x 33 cm |
| For specimen transport | |
| With EPS Insert |