missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Therapak™ Courier Totes
Description
- Shoulder-strap specimen totes constructed of waterproof, scuff-resistant nylon
- Standard 2 in. foldover closure forms secure seal over lid
- Expandable pouch on back of tote holds additional ambient specimens; two document pockets secure requisitions and reports
- Adjustable dividers allow courier to accommodate various shipments
- Available in two sizes with molded, high R-value urethane or EPS foam insert
- Small tote in blue; standard tote in black preprinted with a 2 × 2 1/2 in. red international biohazard symbol
- Meet OSHA and DOT requirements for shipments of biological substances via ground courier transport
- replacement inserts available separately
Specifications
Specifications
| Includes | Urethane insert |
| Certifications/Compliance | Meets OSHA and DOT requirements for shipments of biological substances via ground courier transport |
| Applications | For specimen transport |
| Size | Small |
| Type | With Urethane Insert |
| Dimensions (LxWxH) | 26.03 x 22.2 x 29.2 cm |
| Material | Nylon |