Learn More
Therapak™ Blood-Alcohol Collection and Transport Kit
Catalog No. 22131558
Change view
Click to view available options
Quantity:
24/Cs.
Catalog No. Quantity
22131558 24/Cs.
1 options
Catalog No. 22131558 Supplier Therapak™ Supplier No. 38107
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Includes
Kit includes two 16 x 100mm 10mL gray-top evacuated blood tubes (with Sodium Fluoride-Potassium Oxalate preservative); one blood draw needle and needle holder; an iodine prep pad; a plastic tube holder; two evidence seals; one box seal; one transport bag with absorbent; required box marking labels; and a transport box.
Ideal for use in suspected DUI cases
  • All the necessary components for collection and transport of blood samples from suspected “driving under the influence” offenders
  • Kit contents double sealed in a bag and box
  • Compliant for shipping by ground or air in accordance with Exempt Human Specimen regulations
Specifications
Product Type Blood Alcohol Collection and Transport Kit
For Use With (Application) Collection and transport of Blood samples collected from suspect DUI offenders
Includes Two 16 x 100 mm 10 mL gray top evacuated Blood tubes (with Sodium Fluoride-Potassium Oxalate preservative), one Blood draw needle and needle holder, an Iodine prep pad, Plastic tube holder, two evidence seals, one box seal, one transport bag with absorbent, required box marking labels and a transport box
Quantity 24/Cs.