missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Therapak™ Biological Substance Category B Ambient Shipping Systems
Description
Includes
Each system supplied with absorbent pouch or sheet, cushioning material to secure the inner packaging (except swab canister), biohazard and Biological Substances Category B labels, transport box, a variety of inner packaging choices (depending on the type and and quantity of specimens being transported), list of contents card, and Packing Instructions.
- For transport of blood, urine and other biological substances
- Choice of pre-assembled systems to accommodate a variety of shipping needs
Specifications
Specifications
| Quantity | 1 Ea. |
| Includes | 95kPa Bag with Tube Shuttle |
| Applications | For blood, urine transport |
| Type | Ambient Shipping for Biological Substance Category B |
| Content And Storage | 95kPa Bag; TearTech tube shuttle, cushioning material, biohazard and Biological Substance Category B labels, list of contents card, transport box |
| Capacity | Holds 6 tubes |
| Packaging | Transport box, 95kPa |