Learn More
Therapak™ Biological Substance Category B Ambient Shipping Systems
Include all the necessary marking labels in a convenient format for application at the shipping location
Supplier:  Therapak™ 37925
 View more versions of this product
Catalog No. 22130052
Only null left
Includes: Each system supplied with absorbent pouch or sheet, cushioning material to secure the inner packaging (except swab canister), biohazard and Biological Substances Category B labels, transport box, a variety of inner packaging choices (depending on the type and and quantity of specimens being transported), list of contents card, and Packing Instructions.
Description
Description
  • For transport of blood, urine and other biological substances
  • Choice of pre-assembled systems to accommodate a variety of shipping needs
Specifications
Chemical Identifiers
1 Ea.
Ambient Shipping for Biological Substance Category B
Transport box, 95kPa
Specifications
1 Ea.
Blood, urine transport
Large 95kPa bag; 3 Aqui-Pak absorbent pouch, cushioning material, biohazard, Biological Substance Category B label, list of contents card, and transport box
Transport box, 95kPa
95kPa Large Bag with Aqui-Pak pouch
Ambient Shipping for Biological Substance Category B
Holds 18 tubes
Product Suggestions
Product Suggestions