Learn More
Therapak™ Aqui-Pak™ Absorbent Sleeves
Ideal for Biological Substance and Exempt Human specimen shipments
Supplier: Therapak™ 10314
Description
Ideal for Biological Substance and Exempt Human specimen shipments
- For safe transport of specimen tubes in accordance with DOT/IATA regulations
- Consist of multilayered polymeric material with extremely high absorption properties
- Simultaneously separates and cushions specimen tubes
- Place between primary and secondary container
- Roll it up and place it in a bag or canister
Specifications
| Up to four tubes | |
| Absorbent Sleeve | |
| 150 mL |