Learn More
Therapak™ 95kPa Compliant Canisters
For transport of blood tubes and urine vials and other biological substances
Supplier:  Therapak™ TR23080
 View more versions of this product
Catalog No. 22-130-037
Only null left
Explore available promotions
Description
Description
  • Reusable containers with durable, thick-wall construction to protect specimens
  • Ideal secondary, watertight container
  • Screw cap for secure, watertight seal
  • Small canister constructed of translucent PET to identify specimen leakage; holds up to five tubes
  • Medium canister for harsh transport conditions including dry ice shipments of frozen specimens; holds up to 6 tubes
  • Large canister is most durable of canisters and can be used for biological substances transport between client sites and reference labs; holds up to 20 tubes
Compliance
Compliant with DOT and IATA regulations as a secondary container
Specifications
Specifications
Up to 20 tubes
Comply with DOT and IATA regulations as a secondary container
For transport of biological substances
Large
Product Suggestions
Product Suggestions