Learn More
Therapak™ 95kPa Compliant Canisters
For transport of blood tubes and urine vials and other biological substances
Supplier: Therapak™ 23072
Description
- Reusable containers with durable, thick-wall construction to protect specimens
- Ideal secondary, watertight container
- Screw cap for secure, watertight seal
- Small canister constructed of translucent PET to identify specimen leakage; holds up to five tubes
- Medium canister for harsh transport conditions including dry ice shipments of frozen specimens; holds up to 6 tubes
- Large canister is most durable of canisters and can be used for biological substances transport between client sites and reference labs; holds up to 20 tubes
Compliance
Compliant with DOT and IATA regulations as a secondary container
Specifications
| Up to 12 tubes | |
| Comply with DOT and IATA regulations as a secondary container | |
| For transport of biological substances | |
| Medium |