missing translation for 'onlineSavingsMsg'
Learn More

Therapak™ 24-Hour Urine Containers Prefilled with Preservative Solution

Catalog No. 22130555
Encompass
Change view
Click to view available options
Volume (Metric):
3 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
22130555 3 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22130555 Supplier Therapak™ Supplier No. 38534G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Used to preserve urine samples during collection and transport for subsequent testing and diagnosis of disease

  • Incorporates weak acids into 24-hour containers, alleviating the need for laboratory personnel to fill preservatives into containers on-site or patients to add preservative to the container prior to voiding
  • Choose from two commonly used preservatives for 24-hour collections
  • 3L space-saver size container stores easily onsite or at patient site
  • Collection instructions on each container for patient convenience

Specifications

Product Type 24-Hour Urine Specimen Collection Container
Closure Type Extra-thick Liner Cap
Material Polyethylene
Color Amber
For Use With (Application) Used to preserve Urine samples during collection and transport for subsequent testing and diagnosis of disease
Includes 30 mL Hydrochloric Acid, Plastic cap with extra thick liner
No. per Case 24/Cs.
Volume (Metric) 3 L