missing translation for 'onlineSavingsMsg'
Learn More

Theophylline, Thermo Scientific Chemicals

Catalog No. p-7041775
Change view
Click to view available options
Quantity:
454 g
2.27 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ602035H 454 g
AAJ60203RAT 2.27 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ602035H Supplier Thermo Scientific Chemicals Supplier No. J602035H

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 58-55-9 | C7H8N4O2 | 180.167 g/mol

Theophylline has been used to study its effects on rat gene expression in the ubiquitin-proteasome pathway that regulates spermatogenesis and epididymal sperm quality. Theophylline has also been used as an internal standard for the measurement of plasma paracetamol levels in humans.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Theophylline has been used to study its effects on rat gene expression in the ubiquitin-proteasome pathway that regulates spermatogenesis and epididymal sperm quality. Theophylline has also been used as an internal standard for the measurement of plasma paracetamol levels in humans.

Solubility
Soluble in ethanol (moderately), DMSO, alkali hydroxides, ammonia, dilute hyrochloric acid, nitric acid, dilute aqueous acid, and dilute aqueous base. Insoluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 58-55-9
Molecular Formula C7H8N4O2
Molecular Weight (g/mol) 180.167
MDL Number MFCD00079619
InChI Key ZFXYFBGIUFBOJW-UHFFFAOYSA-N
Synonym theophylline, 1,3-dimethylxanthine, elixophyllin, theophyllin, theolair, nuelin, theophylline anhydrous, respbid, theocin, theo-dur
PubChem CID 2153
ChEBI CHEBI:28177
IUPAC Name 1,3-dimethyl-7H-purine-2,6-dione
SMILES CN1C2=C(C(=O)N(C1=O)C)NC=N2

Specifications

Melting Point 270°C to 274°C
Color White
Odor Odorless
Quantity 454 g
UN Number UN2811
Beilstein 13463
Merck Index 14,9285
Solubility Information Soluble in ethanol (moderately),DMSO,alkali hydroxides,ammonia,dilute hyrochloric acid,nitric acid,dilute aqueous acid,and dilute aqueous base. Insoluble in water.
Formula Weight 180.16
Physical Form Powder
Chemical Name or Material Theophylline
Show More Show Less
Hazard Category H301
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 200-385-7
RTECSNumber XH3850000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only