missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Theophylline-7-acetic Acid 98.0+%, TCI America™
Supplier: TCI America T294125G
Specifications
| Theophylline-7-acetic Acid | |
| 272°C | |
| C9H9N4NaO4 | |
| 25 g | |
| MSFVZSOKOXZSME-UHFFFAOYSA-M | |
| sodium 2-(1,3-dimethyl-2,6-dioxo-2,3,6,7-tetrahydro-1H-purin-7-yl)acetate | |
| 69550 | |
| ≥98.0% (HPLC,T) |
| 652-37-9 | |
| White | |
| MFCD00022832 | |
| acefylline, theophylline-7-acetic acid, acephylline, theophyllineacetic acid, carboxymethyltheophylline, 7-carboxymethyl theophylline, 7-theophyllinylacetic acid, theophyllin-7-ylacetic acid, 7-theophyllinessigsaeure, 7-theophyllineacetic acid | |
| [Na+].CN1C2=C(N(CC([O-])=O)C=N2)C(=O)N(C)C1=O | |
| 260.19 | |
| 238.20 | |
| Crystalline Powder |
Chemical Identifiers
| 652-37-9 | |
| 260.19 | |
| MSFVZSOKOXZSME-UHFFFAOYSA-M | |
| 69550 | |
| [Na+].CN1C2=C(N(CC([O-])=O)C=N2)C(=O)N(C)C1=O |
| C9H9N4NaO4 | |
| MFCD00022832 | |
| acefylline, theophylline-7-acetic acid, acephylline, theophyllineacetic acid, carboxymethyltheophylline, 7-carboxymethyl theophylline, 7-theophyllinylacetic acid, theophyllin-7-ylacetic acid, 7-theophyllinessigsaeure, 7-theophyllineacetic acid | |
| sodium 2-(1,3-dimethyl-2,6-dioxo-2,3,6,7-tetrahydro-1H-purin-7-yl)acetate |
Safety and Handling
RTECSNumber : UO7451300
TSCA : No