Learn More
Theobromine, 99%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General description
• Theobromine is a methylxanthine alkaloid that is a derivative of caffeine
Applications
• Theobromine is a phosphodiesterase inhibitor, weak adenosine receptor antagonist, and smooth muscle relaxant
• As a catabolic product of caffeine, it has antioxidant and prooxidant properties
• It can be used as a standard for quantification of theobromine
Specifications
Specifications
| Melting Point | 290.0°C to 295.0°C |
| Color | White |
| Loss on Drying | 0.5% max. (1 g, 105°C) |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 99% |
| Packaging | Plastic bottle |
| Quantity | 1 kg |
| Beilstein | 26, 2336 |
| Merck Index | 15, 9433 |
| Solubility Information | Solubility in water: slightly soluble. Other solubilities: moderately soluble in ammonia, practically insoluble in benzene, ether, carbon, tetrachloride and chloroform |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Harmful if swallowed. |
| Precautionary Statement | GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Call a POISON CENTER or doctor/physician if you feel unwell. Wash face,hands and any exposed skin thoroughly after handling. |
| EINECSNumber | 201-494-2 |
| RTECSNumber | XH2275000 |
| TSCA | TSCA |
RUO – Research Use Only