Learn More
United Scientific Supplies The Spillnot™
Catalog No. S05903
Change view
Click to view available options
The Spillnot
Catalog No.
S05903 The Spillnot
1 options
Catalog No. S05903 Supplier United Scientific Supplies Supplier No. SPNT01
Uses the principles of Newtonian physics to keep the liquid in the container.
An inexpensive demonstration to intrigue students and challenge their analytical powers.
  • Flexible handle and location of the suspension point above the center of the tray ensure that the forces on the liquid are always directed toward the supporting tray, even when the tray is in motion
  • Sideways forces usually responsible for spills are not transmitted to the tray by the flexible handle
  • Radial forces generated during swinging motion only force the liquid into the container more firmly
Order Info
Dia. x H (excluding handle): 4.37 x 8 in. (11.2 x 20cm). Weight: 5 oz. (139g).
Specifications
Product Type The SpillNot
For Use With (Application) SpillNot™ uses the principles of Newtonian physics to keep the liquid in the container
Weight (English) 0.31 lb.
Weight (Metric) 0.14 kg