Learn More
Decon™ The Pill™ Autoclave Deodorizer
GREENER_CHOICE
Neutralizes odors from autoclaving
$265.10 - $1,170.07
Specifications
Product Type Autoclave Capsule
Color-coded No
Form Capsules
Scent Fresh Scent
Sterility Non-sterile
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
0435523
GREENER_CHOICE
View Documents Promotion Details
Decon™
DC5100
100 Capsules
Pack of 100 Each for $265.10
Case of 20 Pack for $5,302.00
Only null left
0435524
GREENER_CHOICE
View Documents Promotion Details
Decon™
5500
500 Capsules
Pack of 500 Each for $1,170.07
Case of 6 Pack for $7,020.42
Only null left
Description
Description
  • One capsule treats 7 cu. ft. of autoclave capacity
  • Gelatin capsule keeps deodorizer off fingers, ensures economical dose rate
Specifications
Specifications
Autoclave Capsule
Capsules
Non-sterile
Autoclavable
Autoclave Deodorizer
No
Fresh Scent
Clear Amber
Capsules