missing translation for 'onlineSavingsMsg'
Learn More

SOMNI Scientific™ The Keyfill Isoflurane Bottle Adapter

Catalog No. 04777190
Encompass
Change view
Click to view available options
For Use With (Application):
Research Animal Inhalant Anesthesia
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
04777190 Research Animal Inhalant Anesthesia
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04777190 Supplier SOMNI Scientific™ Supplier No. VF6232

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Allows for safe transfer of anesthetic from bottle to key fill vaporizer. The color coded, agent specific keyed block reduces the risk of introducing incorrect agents.

  • Allows for safe transfer of anesthetic from bottle to key fill vaporizer
  • Color coded, agent specific keyed block
  • Reduces the risk of introducing incorrect agents into vaporizer
  • Reduce exposure to Isoflurane

Specifications

For Use With (Application) Research Animal Inhalant Anesthesia
Type Bottle Adapter