missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FlexMAP™ 3D™ Calibration Kit

Catalog No. F3DCALK25
Encompass
Change view
Click to view available options
Quantity:
25 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F3DCALK25 25 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F3DCALK25 Supplier Invitrogen™ Supplier No. F3DCALK25

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The FlexMAP™ 3D™ Calibration Kit

The convenient calibration kit contains xMAP™ calibrators for MagPlex and MicroPlex classification channels and also includes the calibrator for the reporter channel. Lot specific CD is included for importing target values. The kit is intended to be used with the off plate reagent area. Stripwells are included with each kit. Each kit will provide 25 calibrations.

Specifications

Product Line Luminex
Content And Storage Contains 28 strip wells, FM3D Calibration Kit CD, Luminex Classification Calibrator Microspheres, MagPlex Classification Calibrator Microspheres, Luminex Reporter Calibrator Microspheres. Store at 2°C to 8°C.
Quantity 25 reactions
Shipping Condition Wet Ice
For Use With (Equipment) FlexMAP 3D System
Product Type Calibration Kit
How long does it take to acquire ProcartaPlex assay data on the different Luminex instruments?

With everything working smoothly, here is the approximate read time for one plate on different Luminex instruments:

- FLEXMAP 3D Systems and INTELLIFLEX Systems will read 96 well plates in ~20 min, and 384 well plates in ~75 min.
- Luminex 100/200 Systems will read 96 well plates in ~40 min.
- MAGPIX System will read 96 well plates in ~50-60 min. Please note that these are estimates for the read times of the plates only, and do not include time for calibration or set-up of the instrument.

For Research Use Only. Not for use in diagnostic procedures.