missing translation for 'onlineSavingsMsg'
Learn More

3M™ PELTOR™ CH-3 Listen Only Hearing Protector

Catalog No. 19072263
Change view
Click to view available options
Model:
HT52P3E-112
Style:
Hard Hat
Noise Reduction Rating (NRR):
25 dB
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Style Noise Reduction Rating (NRR)
19072263 HT52P3E-112 Hard Hat 25 dB
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19072263 Supplier 3M™ Supplier No. 7100203302

PELTOR™ CH-3 Listen Only Hearing Protector helps workers stay protected in noisy environments while enabling them to hear incoming audio from handheld devices.

  • Hearing protection solution with clear audio reception in noisy environments
  • FLX2 connection jack for easy and reliable connection to a wide assortment of handheld devices
  • Lasting comfort for long duration wear times
  • Designed to withstand harsh environmental elements
  • Audio speakers are optimized for speech frequencies and meet the efficiency demands of many of today's digital devices
  • Bright yellow color for enhanced visibility

Compliance

3M™ ear muffs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.

Specifications

Color Bright Yellow
ANSI S3.1974 Yes
Cushion Color Black
For Use With (Application) Manufacturing
High Visibility Yes
Model HT52P3E-112
Pairs per Package 1 Ea.
Noise Reduction Rating (NRR) 25 dB
Product Type CH-3 Listen Only
Dielectric No
Style Hard Hat
Show More Show Less