Learn More
Analytical Instruments Group (Aig) Thaw-Lyse, 1000X Concentrate, 60ml
Catalog No. 501351696
Encompass
60 milliliter bottle of Thaw-Lyse Buffer, 1000X Concentrate. Thaw-Lyse Buffer is used after thawing samples stored frozen in PROT1 or Smart Tubes. Thaw-Lyse buffer lyses erythrocytes in samples previously frozen in PROT1 or Smart Tubes. This buffer is supplied as a 1000X concentrate. User information must be added to all orders for accurate processing
Catalog No. 501351696 Supplier Analytical Instruments Group (Aig) Supplier No. THAWLYSE1
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
60 milliliter bottle of Thaw-Lyse Buffer, 1000X Concentrate. Thaw-Lyse Buffer is used after thawing samples stored frozen in PROT1 or Smart Tubes. Thaw-Lyse buffer lyses erythrocytes in samples previously frozen in PROT1 or Smart Tubes. This buffer is supplied as a 1000X concentrate. User information must be added to all orders for accurate processing