missing translation for 'onlineSavingsMsg'
Learn More

Thaumatin, TCI America™

$78.29 - $382.13

Chemical Identifiers

CAS 53850-34-3
MDL Number MFCD00132420
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
T1144100MG
View Documents
TCI America
T1144100MG
100 mg
Each for $78.29
Only null left
 
T11441G
View Documents
TCI America
T11441G
1 g
Each for $382.13
Only null left
 
Description

Description

Specifications

Chemical Identifiers

53850-34-3
MFCD00132420

Specifications

53850-34-3
100 mg
Thaumatin
MFCD00132420
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'