missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thallium(III) oxide, 96%
Description
Thallium(III) oxide is used in organic synthesis and as an analytical reagent
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Thallium(III) oxide is used in organic synthesis and as an analytical reagent
Solubility
Insoluble in water and alkalis. Decomposes in HCl and H2SO4
Notes
Keep container tightly closed. Store away from oxidizing agents.
Thallium(III) oxide is used in organic synthesis and as an analytical reagent
Solubility
Insoluble in water and alkalis. Decomposes in HCl and H2SO4
Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications
Specifications
| Boiling Point | 875°C |
| Melting Point | 717°C |
| Quantity | 50 g |
| UN Number | UN1707 |
| Merck Index | 14,9268 |
| Solubility Information | Insoluble in water and alkalis. Decomposes in HCl and H2SO4 |
| Formula Weight | 456.74 |
| Percent Purity | 96% |
| Odor | Odorless |
| Density | 9.65 |
| Show More |
Safety and Handling
| Hazard Category | H300+H330-H373 |
| Hazard Statement | GHS H Statement H300-H330-H373 Fatal if swallowed. Fatal if inhaled. May cause damage to organs through prolonged or repeated exposure. |
| Precautionary Statement | P260-P264b-P270-P271-P284-P301+P310-P304+P340-P310-P314-P330-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: THALLIUM COMPOUNDS, N.O.S. |
| EINECSNumber | 215-229-3 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only