missing translation for 'onlineSavingsMsg'
Learn More

Thallium(III) oxide, 96%

Catalog No. AAA1760109
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1760109 10 g
AAA1760118 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1760109 Supplier Thermo Scientific Chemicals Supplier No. A1760109
Only null left

CAS: 1314-32-5 | O3Tl2 | 456.757 g/mol

Thallium(III) oxide is used in organic synthesis and as an analytical reagent

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thallium(III) oxide is used in organic synthesis and as an analytical reagent

Solubility
Insoluble in water and alkalis. Decomposes in HCl and H2SO4

Notes
Keep container tightly closed. Store away from oxidizing agents.

Chemical Identifiers

CAS 1314-32-5
Molecular Formula O3Tl2
Molecular Weight (g/mol) 456.757
MDL Number MFCD00011276
InChI Key QTQRFJQXXUPYDI-UHFFFAOYSA-N
Synonym oxo oxothallanyloxy thallane, tl2o3, thallium iii oxide, oxo oxothallanyl oxy thallane, thallium oxide ic, thallium iii oxide trace metals basis
PubChem CID 3579754
IUPAC Name oxo(oxothallanyloxy)thallane
SMILES O=[Tl]O[Tl]=O

Specifications

Boiling Point 875°C
Melting Point 717°C
Quantity 10 g
UN Number UN1707
Merck Index 14,9268
Solubility Information Insoluble in water and alkalis. Decomposes in HCl and H2SO4
Formula Weight 456.74
Percent Purity 96%
Odor Odorless
Density 9.65
Chemical Name or Material Thallium(III) oxide
Show More Show Less
Hazard Category H300+H330-H373
Hazard Statement GHS H Statement
H300-H330-H373
Fatal if swallowed.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P260-P264b-P270-P271-P284-P301+P310-P304+P340-P310-P314-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: THALLIUM COMPOUNDS, N.O.S.
EINECSNumber 215-229-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only