missing translation for 'onlineSavingsMsg'
Learn More

Thallium(III) nitrate trihydrate, 99.5% (metals basis)

Catalog No. p-7080396
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA8799906 5 g
AA8799914 25 g
AA8799922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA8799906 Supplier Thermo Scientific Chemicals Supplier No. 08799906
Only null left

CAS: 13453-38-8 | H12N3O15Tl | 498.48 g/mol

Thallium(III) nitrate trihydrate is a reagent used for the rapid oxidation of styrene derivatives to carbonyl compounds, for the regioselective ring expansion of cyclic aralkyl ketones via oxidative rearrangement, for the selective hydrolysis of thioacetals to carbonyl compounds and for a general synthetic method to 19-norsteroids. Mediates ring contraction of cyclic ketones to a series of trans-fused 10-methyl-2-decalones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13453-38-8
Molecular Formula H12N3O15Tl
Molecular Weight (g/mol) 498.48
MDL Number MFCD00149874
InChI Key TWTZVACKNJEUQF-UHFFFAOYSA-N
Synonym nitric acid,thallium 3+ salt, trihydrate 9ci, thallium trinitrate trihydrate, thallium iii nitrate trihydrate
PubChem CID 11270721
SMILES O.O.O.O.O.O.[Tl+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 102°C to 105°C
Physical Form Crystalline
Quantity 5 g
UN Number UN2727
Solubility Information Soluble in water. (95g/L at 20°C).
IUPAC Name thallium(3+);trinitrate;trihydrate
Formula Weight 444.43 (390.38 Anhydrous)
Percent Purity 99.5%
Odor Odorless
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Thallium (III) nitrate trihydrate
Show More Show Less
Hazard Category H272-H300+H330-H373
Hazard Statement GHS H Statement
H300-H330-H272-H373
Fatal if swallowed.
Fatal if inhaled.
May intensify fire
oxidizer.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P210-P220-P221-P260-P264b-P270-P271-P280-P284-P301+P310-P304+P340-P310-P314-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: THALLIUM NITRATE
EINECSNumber 237-325-4
RTECSNumber QV2000000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only