missing translation for 'onlineSavingsMsg'
Learn More

Thallium(I) sulfate, Honeywell Fluka™

Catalog No. p-7072283
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001694 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6001694 Supplier Honeywell Chemicals Supplier No. 88290500G
Only null left

Purum,≥98.0% (RT)

  • ≥95% for general laboratory use.

Chemical Identifiers

CAS 7446-18-6
Molecular Formula O4STl2
Molecular Weight (g/mol) 504.816
MDL Number MFCD00011278
InChI Key YTQVHRVITVLIRD-UHFFFAOYSA-L
Synonym thallous sulfate, thallium sulfate, thallium i sulfate, tharattin, zelio, bonide antzix, th-universal, unii-u9f9qir12t, rcra waste number p115, sulfuric acid, dithallium 1+ salt
PubChem CID 24833
ChEBI CHEBI:81836
IUPAC Name thallium(1+);sulfate
SMILES [O-]S(=O)(=O)[O-].[Tl+].[Tl+]

Specifications

Quantity 500 g
Linear Formula Tl2SO4
UN Number UN1707
Formula Weight 504.83g/mol
Percent Purity ≥98% (RT)
Density 6.77g/mL at 25°C
Chemical Name or Material Thallium(I) sulfate
Hazard Statement P264-P273-P301 + P310-P314
RTECSNumber XG6800000