Learn More
Thallium(I) sulfate, 99.5% min (metals basis)
CAS: 7446-18-6 | O4STl2 | 504.816 g/mol
Supplier: Thermo Scientific Chemicals 04034218
Description
Thallium(I) sulfate is used as a precursor to thallium(I) sulfide, which exhibits high electrical conductivity when exposed to infrared light. It is further used in photocells. It is also mixed with selenium and arsenic to produce low melting glasses.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| (decomposition) | |
| 632°C | |
| Crystalline | |
| 99.5% min (metals basis) | |
| MFCD00011278 | |
| YTQVHRVITVLIRD-UHFFFAOYSA-L | |
| thallium(1+);sulfate | |
| 24833 | |
| 504.8 |
| Thallium(I) sulfate | |
| 7446-18-6 | |
| 50 g | |
| O4STl2 | |
| thallous sulfate, thallium sulfate, thallium i sulfate, tharattin, zelio, bonide antzix, th-universal, unii-u9f9qir12t, rcra waste number p115, sulfuric acid, dithallium 1+ salt | |
| [O-]S(=O)(=O)[O-].[Tl+].[Tl+] | |
| 504.816 | |
| CHEBI:81836 | |
| 6.77 g/mL |
Chemical Identifiers
| 7446-18-6 | |
| 504.816 | |
| YTQVHRVITVLIRD-UHFFFAOYSA-L | |
| 24833 | |
| thallium(1+);sulfate |
| O4STl2 | |
| MFCD00011278 | |
| thallous sulfate, thallium sulfate, thallium i sulfate, tharattin, zelio, bonide antzix, th-universal, unii-u9f9qir12t, rcra waste number p115, sulfuric acid, dithallium 1+ salt | |
| CHEBI:81836 | |
| [O-]S(=O)(=O)[O-].[Tl+].[Tl+] |
Safety and Handling
P260-P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P314-P330-P332+P313-P361-P363-P501c
H300-H311-H315-H319-H372
DOTInformation : Hazard Class: 6.1; Packaging Group: II
EINECSNumber : 231-201-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only