missing translation for 'onlineSavingsMsg'
Learn More

Thallium(I) sulfate, 99.5% min (metals basis)

Catalog No. AA4034209
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4034209 10 g
AA4034218 50 g
AA4034230 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4034209 Supplier Thermo Scientific Chemicals Supplier No. 04034209

CAS: 7446-18-6 | O4STl2 | 504.816 g/mol

Thallium(I) sulfate is used as a precursor to thallium(I) sulfide, which exhibits high electrical conductivity when exposed to infrared light. It is further used in photocells. It is also mixed with selenium and arsenic to produce low melting glasses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7446-18-6
Molecular Formula O4STl2
Molecular Weight (g/mol) 504.816
MDL Number MFCD00011278
InChI Key YTQVHRVITVLIRD-UHFFFAOYSA-L
Synonym thallous sulfate, thallium sulfate, thallium i sulfate, tharattin, zelio, bonide antzix, th-universal, unii-u9f9qir12t, rcra waste number p115, sulfuric acid, dithallium 1+ salt
PubChem CID 24833
ChEBI CHEBI:81836
IUPAC Name thallium(1+);sulfate
SMILES [O-]S(=O)(=O)[O-].[Tl+].[Tl+]

Specifications

Boiling Point (decomposition)
Melting Point 632°C
Physical Form Crystalline
Quantity 10 g
Assay Percent Range 99.5% min (metals basis)
Formula Weight 504.8
Density 6.77
Chemical Name or Material Thallium(I) sulfate
Hazard Category H300-H311-H315-H319-H372
Precautionary Statement P260-P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P314-P330-P332+P313-P361-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 231-201-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only