missing translation for 'onlineSavingsMsg'
Learn More

Thallium(I) iodide, Puratronic™, 99.9995% (metals basis)

Catalog No. AA4300118
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4300118 50 g
AA4300109 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4300118 Supplier Thermo Scientific Chemicals Supplier No. 04300118

CAS: 7790-30-9 | ITl | 331.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7790-30-9
Molecular Formula ITl
Molecular Weight (g/mol) 331.28
MDL Number MFCD00011279
InChI Key CMJCEVKJYRZMIA-UHFFFAOYSA-M
Synonym thallium iii iodide, thallium iodide tli2, thallium iodide ous, thallium i iodide, anhydrous, ampuled under argon trace metals basis 10g, tli
IUPAC Name λ¹-thallanylium iodide
SMILES [I-].[Tl+]

Specifications

Boiling Point 824°C
Melting Point 440°C
Color Yellow
Physical Form Powder
Quantity 50 g
UN Number UN1707
Merck Index 14,9264
Formula Weight 331.27
Percent Purity 99.9995%
Grade Puratronic
Odor Odorless
Sensitivity Light sensitive
Density 7.1 g/mL
Assay (metals basis)
Chemical Name or Material Thallium (I) iodide
Show More Show Less
Hazard Category H300+H330-H373
Hazard Statement GHS H Statement
H300-H330-H373
Fatal if swallowed.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P260-P264b-P270-P271-P284-P301+P310-P304+P340-P310-P314-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: THALLIUM COMPOUNDS, N.O.S.
EINECSNumber 232-199-7
RTECSNumber XG5425000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only