missing translation for 'onlineSavingsMsg'
Learn More

Thallium(I) hexafluorophosphate(V), 97% min, Thermo Scientific Chemicals

Catalog No. p-7050881
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3962706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA3962706 Supplier Thermo Scientific Chemicals Supplier No. 3962706

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 60969-19-9 | Molecular Formula: F6PTl | Molecular Weight: 349.344 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 60969-19-9
Molecular Formula F6PTl
Molecular Weight (g/mol) 349.344
MDL Number MFCD00049807
InChI Key FRZBCOUMLHRKRT-UHFFFAOYSA-N
Synonym thallium hexafluorophosphate, tlpf6, thallium i hexafluorophosphate, thallium 1+ hexafluorophosphate, thallium 1+ hexafluoro-$l^ 5-phosphanuide, $l^ 1-thallanylium hexafluoro-$l^ 5-phosphanuide
PubChem CID 10904204
IUPAC Name thallium(1+);hexafluorophosphate
SMILES F[P-](F)(F)(F)(F)F.[Tl+]

Specifications

Physical Form Crystalline
Quantity 5 g
UN Number UN1707
Solubility Information Soluble in water.
Formula Weight 349.34
Percent Purity ≤97%
Chemical Name or Material Thallium(I) hexafluorophosphate(V)
Hazard Category H300+H330-H373
Hazard Statement GHS H Statement
H300-H330-H373
Fatal if swallowed.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P260-P264b-P270-P271-P284-P301+P310-P304+P340-P310-P314-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: THALLIUM COMPOUNDS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only