Learn More
Thallic trifluoroacetate, tech., Thermo Scientific Chemicals
$480.76 - $480.76
Chemical Identifiers
| CAS | 23586-53-0 |
|---|---|
| Molecular Formula | C6F9O6Tl |
| Molecular Weight (g/mol) | 543.42 |
| MDL Number | MFCD00000414 |
| InChI Key | PSHNNUKOUQCMSG-UHFFFAOYSA-K |
| Synonym | thallium iii trifluoroacetate, acetic acid, trifluoro-, thallium 3+ salt, acetic acid,2,2,2-trifluoro-, thallium 3+ salt 3:1, thallium 3+ ; 2,2,2-trifluoroacetate, thallium tris trifluoroacetate, trifluoroacetic acid thallium iii, thallic trifluoroacetate, tech, thallium 3+ tritrifluoroacetate |
| PubChem CID | 90200 |
| IUPAC Name | thallium(3+);2,2,2-trifluoroacetate |
| SMILES | C(=O)(C(F)(F)F)[O-].C(=O)(C(F)(F)F)[O-].C(=O)(C(F)(F)F)[O-].[Tl+3] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC420760250
|
Thermo Scientific Chemicals
420760250 |
25 g | Glass bottle |
Each for $480.76
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 23586-53-0 | |
| 543.42 | |
| PSHNNUKOUQCMSG-UHFFFAOYSA-K | |
| 90200 | |
| C(=O)(C(F)(F)F)[O-].C(=O)(C(F)(F)F)[O-].C(=O)(C(F)(F)F)[O-].[Tl+3] |
| C6F9O6Tl | |
| MFCD00000414 | |
| thallium iii trifluoroacetate, acetic acid, trifluoro-, thallium 3+ salt, acetic acid,2,2,2-trifluoro-, thallium 3+ salt 3:1, thallium 3+ ; 2,2,2-trifluoroacetate, thallium tris trifluoroacetate, trifluoroacetic acid thallium iii, thallic trifluoroacetate, tech, thallium 3+ tritrifluoroacetate | |
| thallium(3+);2,2,2-trifluoroacetate |
Specifications
| Beige | |
| Fine Powder | |
| C6F9O6Tl | |
| MFCD00000414 | |
| 03,286; 04,498; 05,658; 06,579; 07,365; 08,478; 10,397; 05,654 | |
| PSHNNUKOUQCMSG-UHFFFAOYSA-K | |
| thallium(3+);2,2,2-trifluoroacetate | |
| 90200 | |
| Technical | |
| Glass bottle |
| 23586-53-0 | |
| 25 g | |
| (CF3COO)3Tl | |
| 02, II, 186 | |
| thallium iii trifluoroacetate, acetic acid, trifluoro-, thallium 3+ salt, acetic acid,2,2,2-trifluoro-, thallium 3+ salt 3:1, thallium 3+ ; 2,2,2-trifluoroacetate, thallium tris trifluoroacetate, trifluoroacetic acid thallium iii, thallic trifluoroacetate, tech, thallium 3+ tritrifluoroacetate | |
| C(=O)(C(F)(F)F)[O-].C(=O)(C(F)(F)F)[O-].C(=O)(C(F)(F)F)[O-].[Tl+3] | |
| 543.42 | |
| 543.42 | |
| Authentic | |
| Thallic trifluoroacetate |
Safety and Handling
GHS H Statement:
Fatal if swallowed.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Toxic to aquatic life with long lasting effects.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 245-761-1
RUO – Research Use Only