missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thallic trifluoroacetate, tech., Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Beige |
| Physical Form | Fine Powder |
| Quantity | 25 g |
| Linear Formula | (CF3COO)3Tl |
| Beilstein | 02, II, 186 |
| Fieser | 03,286; 04,498; 05,658; 06,579; 07,365; 08,478; 10,397; 05,654 |
| Formula Weight | 543.42 |
| Grade | Technical |
| Infrared Spectrum | Authentic |
| Packaging | Glass bottle |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement: Fatal if swallowed. Fatal if inhaled. May cause damage to organs through prolonged or repeated exposure. Toxic to aquatic life with long lasting effects. |
| Precautionary Statement | GHS P Statement: IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Immediately call a POISON CENTER or doctor/physician. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. Do not breathe dust/fume/gas/mist/vapors/spray. Avoid release to the environment. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
| EINECSNumber | 245-761-1 |
RUO – Research Use Only