missing translation for 'onlineSavingsMsg'
Learn More

Thallic acetate, 98%, Thermo Scientific™

$1988.61 - $1988.61

Chemical Identifiers

CAS 2570-63-0
Molecular Formula C6H9O6Tl
Molecular Weight (g/mol) 381.51
MDL Number MFCD00008698
Synonym Acetic acid, thallium(III) salt
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC420751000
SDS View Documents
Thermo Scientific Chemicals
420751000
100 g Glass bottle
Each for $1,988.61
Only null left
 
Description

Description

Specifications

Chemical Identifiers

2570-63-0
381.51
Acetic acid, thallium(III) salt
C6H9O6Tl
MFCD00008698

Specifications

182°C
White
100 g
C6H9O6Tl
MFCD00008698
01,1150; 02,406; 03,286; 04,492; 05,655; 07,360
Acetic acid, thallium(III) salt
381.51
98%
Glass bottle
2570-63-0
Crystalline Powder
98%
(CH3CO2)3Tl
02, 96
15, 9406
Solubility in water: decomposes
381.51
Authentic
Thallic acetate, 98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Fatal if swallowed.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Toxic to aquatic life with long lasting effects.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 219-913-2

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only