Learn More
Texwipe™ Replacement Covers for AlphaMop™ Cleanroom Mops
Suitable for requirements of Class 3-5 environments
Supplier:  Texwipe™ STX7118
Catalog No. 19-139-878
Only null left
Explore available promotions
Includes: Lot-specific certificates of compliance and irradiation
Description
Description
  • Made of pre-sterilized double-knit AlphaWipe™ polyester
  • Gamma irradiated; covers and pads double-bagged together for sterlity assurance
  • For use in controlled, aseptic and sterile environments
  • Mopping walls, floors and ceilings
  • Applying and removing cleaning and disinfecting solutions
  • Extremely low levels of releasable fibers
  • Featured fitting compatibility with lat-padded head mop easily changeable, laundered polyester
  • Completely independent quality control audits for sterility assurance
  • Easy-to-read lot code and expiration date on each bag
  • Eliminates preparation, documentation and validation of autoclaving covers and pads
  • Conforms to textured floors and other surfaces
Compliance
Sterility Assurance Level (SAL) of 106 in accordance with ANSI™/AAMI™/ISO™11137
Specifications
Specifications
Autoclavable
Product Suggestions
Product Suggestions