Learn More
Texwipe™ IdealMat™ Contamination Control Mats - 60 Layer
Apply directly to floor
Supplier:  Texwipe™ AMA364682B
 View more versions of this product
Catalog No. 19-166-876
Only null left
Explore available promotions
Description
Description
Apply directly to floor
Choose 30- or 60-layer mats.
  • Consistent adhesive coating on each layer captures particulates from foot traffic and equipment wheels
  • Antimicrobial agent in coating reduces potential for spreading germs and bacteria into controlled area
Specifications
Specifications
46 x 36 in.
Blue
60 Sheets/Mat
Product Suggestions
Product Suggestions