Learn More
Texwipe™ IdealMat™ Contamination Control Mats - 60 Layer
Apply directly to floor
Supplier: Texwipe™ AMA254582B
Description
Apply directly to floor
Choose 30- or 60-layer mats.
- Consistent adhesive coating on each layer captures particulates from foot traffic and equipment wheels
- Antimicrobial agent in coating reduces potential for spreading germs and bacteria into controlled area
Specifications
| 45 x 25 in. | |
| Blue | |
| 60 Sheets/Mat |